Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzyl benzoate, ≥98% (GC), MilliporeSigma™ Supelco™

Catalog No. 111019355 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9355 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9355 Supplier MilliporeSigma Supelco Supplier No. 681831ML
Add to Cart
Add to Cart

Benzyl benzoate may undergo hydrogenation reaction catalyzed by a system of [Ru(acac)3](Where Ru-ruthenium/acac-acetylacetonate) with MeC(CH2PPh2)3(tripodal triphosphine ligand) and in the presence of excess of triethylamine in trifluoroethanol to obtain benzyl alcohol.

Featured Industries: Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 120-51-4
Ignition Point 896°F
Vapor Pressure 1 mmHg (125°C)
Density 1.118 g/mL (at 20°C)
Flash Point 158°C - closed cup
Physical Form Neat
Linear Formula C6H5 COOCH2C6H5
MDL Number MFCD00003075
Melting Point 17°C to 20°C
Boiling Point 323°C to 324°C
Synonym Benzoic acid benzyl ester
Type Analytical Standard
Quantity 1 mL
Formula Weight 212.24
Percent Purity ≥98% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.