Learn More
Medchemexpress LLC Benzyl benzoate-d12 | 352431-26-6 | 99.9% | 5 MG
SDP
Catalog No. 5000408369
Encompass_Preferred
Benzyl benzoate-d12 is a deuterium-labeled form of Benzyl benzoate, also known as Benzoic acid benzyl ester-d12. It serves as a fragrance ingredient in cosmetic products and is utilized in research for Scabies and Demodex-associated inflammatory skin conditions. It is for research use only and not sold to patients. Deuteration, the incorporation of stable heavy isotopes of hydrogen, carbon, and other elements, is a key feature, offering potential benefits by affecting the pharmacokinetic and metabolic profiles of drugs during development.
  • Used as a fragrance ingredient in cosmetic products.
  • Utilized in research for Scabies and Demodex-associated inflammatory skin conditions.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration affects the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-08369 Supplier Medchemexpress LLC Supplier No. HYB0935S15MG
Benzyl benzoate-d12 is a deuterium-labeled form of Benzyl benzoate, also known as Benzoic acid benzyl ester-d12. It serves as a fragrance ingredient in cosmetic products and is utilized in research for Scabies and Demodex-associated inflammatory skin conditions. It is for research use only and not sold to patients. Deuteration, the incorporation of stable heavy isotopes of hydrogen, carbon, and other elements, is a key feature, offering potential benefits by affecting the pharmacokinetic and metabolic profiles of drugs during development.
  • Used as a fragrance ingredient in cosmetic products.
  • Utilized in research for Scabies and Demodex-associated inflammatory skin conditions.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration affects the pharmacokinetic and metabolic profiles of drugs.