Learn More
Medchemexpress LLC Benzyl benzoate-d12 | 352431-26-6 | MFCD01317469 | 99.9% | 224.32 g/mol | C14D12O2 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYB0935S110MG
Encompass_Preferred
Benzyl benzoate-d12 is a deuterium-labeled benzyl benzoate supplied as a high-purity stable isotope standard for analytical and research applications. It is intended for use as an internal standard in mass spectrometry, tracer studies, and quantitative analyses, and should be stored under recommended refrigerated or frozen conditions for long-term stability.
  • High purity (99.85%) suitable for analytical use.
  • Deuterium-labeled internal standard for mass spectrometry.
  • Molecular weight 224.32 g/mol and formula C14D12O2.
  • Supplied as a 10 mg vial for laboratory use.
  • Certificate of analysis and safety data sheet available.
Catalog No. 50-002-96699