Learn More
Medchemexpress LLC Benzyl benzoate-d5 | 347840-01-1 | 99.8% | 217.27 | C14H7D5O2 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYB0935S10MG
Encompass_Preferred
Benzyl benzoate-d5 is a deuterium-labeled analogue of benzyl benzoate supplied as a high-purity analytical standard for research use. It is intended for use as an internal standard or reference in mass spectrometry and other analytical methods requiring isotopically labeled compounds, and is provided as a pre-weighed solid suitable for laboratory applications.
  • Deuterium-labeled internal standard for analytical assays.
  • High purity (99.8%).
  • Molecular weight 217.27 and molecular formula C14H7D5O2.
  • Available in multiple small pack sizes for method development and validation.
Catalog No. 50-002-96745