Learn More
Medchemexpress LLC Benzyl benzoate-d5 | 347840-01-1 | 99.8% | 25 MG
SDP
Catalog No. 5000450482
Encompass_Preferred
Benzyl benzoate-d5 is the deuterium-labeled version of Benzyl benzoate (Benzoic acid benzyl ester), a fragrance ingredient in cosmetic products. It is used in research for conditions such as Scabies and Demodex-associated inflammatory skin conditions. Stable heavy isotopes, such as deuterium, have been incorporated into drug molecules as tracers for quantification during drug development, with potential to affect pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-50482 Supplier Medchemexpress LLC Supplier No. 25MGHYB0935S
Benzyl benzoate-d5 is the deuterium-labeled version of Benzyl benzoate (Benzoic acid benzyl ester), a fragrance ingredient in cosmetic products. It is used in research for conditions such as Scabies and Demodex-associated inflammatory skin conditions. Stable heavy isotopes, such as deuterium, have been incorporated into drug molecules as tracers for quantification during drug development, with potential to affect pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS