Learn More
Medchemexpress LLC Benzyl benzoate-d5 | 347840-01-1 | 99.8% | 50 MG
SDP
Catalog No. 5000408950
Encompass_Preferred
Benzyl benzoate-d5 is a deuterium-labeled form of Benzyl benzoate. Benzyl benzoate (benzoic acid benzyl ester) is used as a fragrance ingredient in cosmetic products and in research related to scabies and demodex-associated inflammatory skin conditions. It is for research use only.
  • Deuterium-labeled compound
  • Can be used as a tracer
  • Functions as an internal standard for quantitative analysis
  • Suitable for NMR, GC-MS, or LC-MS techniques
  • Potential to influence pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-08950 Supplier Medchemexpress LLC Supplier No. HYB0935S50MG
Benzyl benzoate-d5 is a deuterium-labeled form of Benzyl benzoate. Benzyl benzoate (benzoic acid benzyl ester) is used as a fragrance ingredient in cosmetic products and in research related to scabies and demodex-associated inflammatory skin conditions. It is for research use only.
  • Deuterium-labeled compound
  • Can be used as a tracer
  • Functions as an internal standard for quantitative analysis
  • Suitable for NMR, GC-MS, or LC-MS techniques
  • Potential to influence pharmacokinetic and metabolic profiles of drugs