Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Benzyl bromide

Catalog No. B17905100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B17905100G 100 g
B1790525KG 2.5 kg
B1790525G 25 g
B17905500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. B17905100G Supplier Sigma Aldrich Supplier No. B17905100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Benzyl bromide

Specifications

CAS 100-39-0
Melting Point -3°C to +1°C (lit.)
Density 1.438 g/mL (at 25°C (literature))
Boiling Point 198°C to 199°C (lit.)
Molecular Formula C7H7Br
Refractive Index n20/D 1.575 (literature)
Linear Formula C6H5CH2Br
MDL Number MFCD00000172
Quantity 100 g
Synonym alpha-Bromotoluene
Molecular Weight (g/mol) 171.03
Percent Purity 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.