Learn More
Medchemexpress LLC Benzyl butyl phthalate | 85-68-7 | 312.36 | 100 MG
SDP
Catalog No. 5000455069
Encompass_Preferred
Benzyl butyl phthalate, a member of phthalic acid esters (PAEs), is known to trigger the migration and invasion of hemangioma (HA) cells by upregulating Zeb1. It also activates the aryl hydrocarbon receptor (AhR) in breast cancer cells, stimulating SPHK1/S1P/S1PR3 signaling and enhancing the formation of metastasis-initiating breast cancer stem cells (BCSCs).
  • Member of phthalic acid esters (PAEs)
  • Triggers migration and invasion of hemangioma (HA) cells via Zeb1 upregulation
  • Activates aryl hydrocarbon receptor (AhR) in breast cancer cells
  • Stimulates SPHK1/S1P/S1PR3 signaling
  • Enhances formation of metastasis-initiating breast cancer stem cells (BCSCs)
Catalog No. 50-004-55069 Supplier Medchemexpress LLC Supplier No. HYW011338100MG
Benzyl butyl phthalate, a member of phthalic acid esters (PAEs), is known to trigger the migration and invasion of hemangioma (HA) cells by upregulating Zeb1. It also activates the aryl hydrocarbon receptor (AhR) in breast cancer cells, stimulating SPHK1/S1P/S1PR3 signaling and enhancing the formation of metastasis-initiating breast cancer stem cells (BCSCs).
  • Member of phthalic acid esters (PAEs)
  • Triggers migration and invasion of hemangioma (HA) cells via Zeb1 upregulation
  • Activates aryl hydrocarbon receptor (AhR) in breast cancer cells
  • Stimulates SPHK1/S1P/S1PR3 signaling
  • Enhances formation of metastasis-initiating breast cancer stem cells (BCSCs)