Learn More
Medchemexpress LLC Benzyl butyl phthalate-d4 | 93951-88-3 | 98.38% | 316.38 | 5 MG
SDP
Catalog No. 5000454903
Encompass_Preferred
Benzyl butyl phthalate-d4 is the deuterium-labeled version of Benzyl butyl phthalate. Benzyl butyl phthalate, a member of phthalic acid esters (PAEs), has been shown to induce the migration and invasion of hemangioma (HA) cells by upregulating Zeb1. Additionally, it activates the aryl hydrocarbon receptor (AhR) in breast cancer cells, which stimulates SPHK1/S1P/S1PR3 signaling and promotes the formation of metastasis-initiating breast cancer stem cells (BCSCs). This compound is intended for research use only.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuterium labeled compound
  • Potential to affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-54903 Supplier Medchemexpress LLC Supplier No. HYW011338S5MG
Benzyl butyl phthalate-d4 is the deuterium-labeled version of Benzyl butyl phthalate. Benzyl butyl phthalate, a member of phthalic acid esters (PAEs), has been shown to induce the migration and invasion of hemangioma (HA) cells by upregulating Zeb1. Additionally, it activates the aryl hydrocarbon receptor (AhR) in breast cancer cells, which stimulates SPHK1/S1P/S1PR3 signaling and promotes the formation of metastasis-initiating breast cancer stem cells (BCSCs). This compound is intended for research use only.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuterium labeled compound
  • Potential to affect pharmacokinetic and metabolic profiles of drugs