Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzyl butyl phthalate, MilliporeSigma™ Supelco™

Catalog No. 111019522 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9522 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9522 Supplier MilliporeSigma Supelco Supplier No. 36927250MG
Add to Cart
Add to Cart
Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

Ignition Point 450°F
CAS 85-68-7
Vapor Pressure 0.16 mmHg (150°C)
Vapor Density 10.8 (vs. air)
Density 1.1 g/mL (at 25°C)
Flash Point 113.0°C - Closed Cup
Physical Form Neat
Linear Formula 2-[CH3(CH2)3O2 C]C6H4 CO2 CH2C6H5
MDL Number MFCD00009440
Type Analytical Standard
Quantity 250 mg
Formula Weight 312.36
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.