Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzyl Chloride Solution, 1000μg/mL in hexane, Ultra Scientific

Catalog No. USEPA1196
Encompass
Change view
Click to view available options
Quantity:
1 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
US-EPA-1196 1 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. US-EPA-1196 Supplier Agilent Technologies Supplier No. EPA11961
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 100-44-7 , 110-54-3
Molecular Formula C7H7Cl
Molecular Weight (g/mol) 126.583
InChI Key KCXMKQUNVWSEMD-UHFFFAOYSA-N
PubChem CID 7503
ChEBI CHEBI:615597
IUPAC Name chloromethylbenzene
SMILES C1=CC=C(C=C1)CCl

Specifications

CAS Max % 99.84, 0.15
Analyte or Component Names n-hexane,benzal chloride
Element n-hexane, Benzyl Chloride
Solvent or Matrix n-hexane
Quantity 1 x 1 mL
Concentration or Composition (by Analyte or Components) 1000μg/mL
Chemical Name or Material Benzyl Chloride Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.