Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzyl chloroformate, 95%, stab. with ca 0.1% sodium carbonate

Catalog No. AAA1568218
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1568218 50 g
AAA1568230 250 g
AAA156820B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1568218 Supplier Thermo Scientific Chemicals Supplier No. A1568218
  • $79.70
    $64.65 / Each of 1

    Save $15.05

    19% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 501-53-1 | C8H7ClO2 | 170.592 g/mol

Benzyl chloroformate is widely used as a reactive chemical intermediate in plastic, pharmaceutical, agricultural and organic chemicals. It is useful for the introduction of carboxybenzyl (cbz) protecting group for amines such as aniline in organic synthesis. It is also involved in the synthesis of 1,2,4-oxadiazoles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzyl chloroformate is widely used as a reactive chemical intermediate in plastic, pharmaceutical, agricultural and organic chemicals. It is useful for the introduction of carboxybenzyl (cbz) protecting group for amines such as aniline in organic synthesis. It is also involved in the synthesis of 1,2,4-oxadiazoles.

Solubility
Miscible with ether, acetone and benzene.

Notes
Store in cool place. Moisture sensitive.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 501-53-1
Molecular Formula C8H7ClO2
Molecular Weight (g/mol) 170.592
MDL Number MFCD00000640
InChI Key HSDAJNMJOMSNEV-UHFFFAOYSA-N
Synonym benzyl chloroformate, carbobenzoxy chloride, benzyl chlorocarbonate, cbz chloride, benzyloxycarbonyl chloride, carbobenzyloxy chloride, carbonochloridic acid, phenylmethyl ester, benzylcarbonyl chloride, cbz-cl, chloroformic acid, benzyl ester
PubChem CID 10387
IUPAC Name benzyl carbonochloridate
SMILES C1=CC=C(C=C1)COC(=O)Cl

Specifications

Melting Point -20°C
Density 1.195
Boiling Point 178°C to 180°C
Flash Point 125°C (257°F)
Odor Acrid
Refractive Index 1.517
Quantity 50 g
UN Number UN1739
Sensitivity Moisture sensitive
Merck Index 14,1801
Solubility Information Miscible with ether,acetone and benzene.
Formula Weight 170.6
Percent Purity 95%
Chemical Name or Material Benzyl chloroformate, stabilized with ≈0.1% sodium carbonate
Show More Show Less
Hazard Category H227-H290-H303-H314-H332-H335-H336-H350-H373
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P201-P202-P210-P234-P235-P260-P264b-P271-P281-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P390-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: I; Proper Shipping Name: BENZYL CHLOROFORMATE
EINECSNumber 207-925-0
RTECSNumber LQ5860000
TSCA Yes
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.