Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzyl chloroformate, Thermo Scientific™

Catalog No. AC124490000
Catalog No. AC124490000 Supplier Thermo Scientific Chemicals Supplier No. 124490000

missing translation for 'validMainframeMsg'

Coupling reagent

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 501-53-1 , 108-88-3 , 100-44-7
Molecular Formula C8H7ClO2
Molecular Weight (g/mol) 170.592
InChI Key HSDAJNMJOMSNEV-UHFFFAOYSA-N
Synonym benzyl chloroformate, carbobenzoxy chloride, benzyl chlorocarbonate, cbz chloride, benzyloxycarbonyl chloride, carbobenzyloxy chloride, carbonochloridic acid, phenylmethyl ester, benzylcarbonyl chloride, cbz-cl, chloroformic acid, benzyl ester
PubChem CID 10387
IUPAC Name benzyl carbonochloridate
SMILES C1=CC=C(C=C1)COC(=O)Cl

Specifications

Linear Formula C6H5CH2OOCCl
Density 1.0100g/mL
Solubility Information Solubility in water: decomposes.
Formula Weight 170.6
Percent Purity 45 to 53% (w/w)(GC)
Specific Gravity 1.01
Flash Point 7°C
Chemical Name or Material Benzyl chloroformate
Hazard Category Danger
Hazard Statement GHS H Statement May cause drowsiness or dizziness. May cause cancer. May be fatal if swallowed and enters airways. May cause damage to organs through prolonged or repeated exposure. Very toxic to aquatic life with long lasting effects. Causes severe skin burns and eye damage. Suspected of damaging the unborn child. Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Do not breathe dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician. Avoid release to the environment.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.