Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzyl cinnamate, 99%

Catalog No. AC215291000
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC215291000 100 g Glass bottle
AC215290050 5 g Glass bottle
AC215295000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC215291000 Supplier Thermo Scientific Chemicals Supplier No. 215291000
Only null left
Add to Cart

CAS: 103-41-3 | C16H14O2 | 238.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103-41-3
Molecular Formula C16H14O2
Molecular Weight (g/mol) 238.29
MDL Number MFCD00004789,MFCD00004789,MFCD00004789
InChI Key NGHOLYJTSCBCGC-VAWYXSNFSA-N
Synonym benzyl alcohol cinnamic ester, benzylcinnamate, benzyl alcohol, cinnamate, wln: r1u1vo1r, benzyl .gamma.-phenylacrylate, benzyl cis-cinnamate, benzyl z-cinnamate, 2-propenoic acid, 3-phenyl-, phenylmethyl ester, z-benzyl 3-phenylacrylate, cinnamic acid, benzyl ester, z
PubChem CID 15558051
IUPAC Name benzyl (Z)-3-phenylprop-2-enoate
SMILES O=C(OCC1=CC=CC=C1)\C=C\C1=CC=CC=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 33°C to 37°C
Color Colorless to Yellow
Boiling Point 195°C to 200°C (5.0 mmHg)
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4025 to 1.4045
Linear Formula C6H5CH=CHCO2CH2C6H5
Quantity 100 g
Beilstein 09, 584
Merck Index 15, 1133
Solubility Information Solubility in water: practically insoluble. Other solubilities: soluble in alcohol,ether and oils,practically insoluble in propylene glycol and,glycerin
Formula Weight 238.29
Percent Purity 99%
Physical Form Crystalline Mass or Liquid After Melting
Chemical Name or Material Benzyl cinnamate, 99%
Show More Show Less
EINECSNumber 203-109-3
RTECSNumber GD8400000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.