Learn More
Medchemexpress LLC Benzyl cinnamate-d5 | 347840-02-2 | 10 MG
SDP
Catalog No. 5000420161
Encompass_Preferred
Benzyl cinnamate-d5 is a deuterium-labeled derivative of Benzyl cinnamate, which is naturally found in Balsam of Peru, Tolu balsam, Sumatra and Penang benzoin, and is a primary component of copaiba balsam. It is utilized in strong oriental perfumes and as a fixative. Deuteration of drug molecules has gained attention due to its potential impact on pharmacokinetic and metabolic profiles.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Potential to impact pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-20161 Supplier Medchemexpress LLC Supplier No. HYN7090S10MG
Benzyl cinnamate-d5 is a deuterium-labeled derivative of Benzyl cinnamate, which is naturally found in Balsam of Peru, Tolu balsam, Sumatra and Penang benzoin, and is a primary component of copaiba balsam. It is utilized in strong oriental perfumes and as a fixative. Deuteration of drug molecules has gained attention due to its potential impact on pharmacokinetic and metabolic profiles.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Potential to impact pharmacokinetic and metabolic profiles of drugs