Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Benzyl cinnamate-d5 | 347840-02-2 | MFCD01074144 | 1 MG
SDP

Catalog No. 5000419686
Encompass_Preferred

Benzyl cinnamate-d5 is a chemical compound with the molecular formula C₁₆H₉D₅O₂ and a molecular weight of 243.31. It appears as a white to off-white solid-liquid mixture. This product has been tested and complies with specifications, showing a purity of 98.15% by HPLC and an isotopic enrichment of 99.00%. It is intended for research use only and has not been fully validated for medical applications.

  • Store at 4°C, sealed, away from moisture and light.
  • For solutions, store at -80°C for 6 months or -20°C for 1 month, sealed, away from moisture and light.
  • Purity (HPLC) of 98.15%.
  • Isotopic enrichment of 99.00%.

Catalog No. 50-004-19686 Supplier Medchemexpress LLC Supplier No. HYN7090S1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Benzyl cinnamate-d5 is a chemical compound with the molecular formula C₁₆H₉D₅O₂ and a molecular weight of 243.31. It appears as a white to off-white solid-liquid mixture. This product has been tested and complies with specifications, showing a purity of 98.15% by HPLC and an isotopic enrichment of 99.00%. It is intended for research use only and has not been fully validated for medical applications.

  • Store at 4°C, sealed, away from moisture and light.
  • For solutions, store at -80°C for 6 months or -20°C for 1 month, sealed, away from moisture and light.
  • Purity (HPLC) of 98.15%.
  • Isotopic enrichment of 99.00%.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.