Learn More
eMolecules​ BENZYL CIS-3-HYDROXYCYCLOBUTANECARBOXYLATE | AstaTech | 552849-32-8 | MFCD20441969 | 206.241 | C12H14O3 | 95.000 | O[C@H]1C[C@H](C1)C(=O)OCc1ccccc1 | 1g | 449778529
Supplier:  eMolecules​ 449778529
Encompass_Preferred
BENZYL CIS-3-HYDROXYCYCLOBUTANECARBOXYLATE | AstaTech | 552849-32-8 | MFCD20441969 | 206.241 | C12H14O3 | 95.000 | O[C@H]1C[C@H](C1)C(=O)OCc1ccccc1 | 1g | 449778529
Catalog No. NC2235655