Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzyl D-(-)-Mandelate 98.0+%, TCI America™
SDP

Catalog No. M13545G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M13545G 5 g
M135425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M13545G Supplier TCI America Supplier No. M13545G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 97415-09-3
Molecular Formula C15H14O3
Molecular Weight (g/mol) 242.274
MDL Number MFCD00674031
InChI Key JFKWZVQEMSKSBU-CQSZACIVSA-N
Synonym D-(-)-Mandelic Acid Benzyl Ester
PubChem CID 853755
IUPAC Name benzyl (2R)-2-hydroxy-2-phenylacetate
SMILES C1=CC=C(C=C1)COC(=O)C(C2=CC=CC=C2)O

Specifications

Melting Point 107°C
Color White-Yellow
Quantity 5 g
Formula Weight 242.27
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Benzyl D-(-)-Mandelate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.