Learn More
Medchemexpress LLC Benzyl DC-81 | 127810-79-1 | C20H20N2O3 | 5 MG
SDP
Catalog No. 5000372703
Encompass_Preferred
Benzyl DC-81 (Compound 6a) is an anticancer agent with antiproliferative activity against A375 and MCF-7 cells. It has an IC50 of 1 μM against CH1 human ovarian cells and 3 μM against L1210 Leukemia cells. This product is intended for research use only.
  • Anticancer agent
  • Antiproliferative activity against A375 and MCF-7 cells
  • IC50 of 1 μM against CH1 human ovarian cells
  • IC50 of 3 μM against L1210 Leukemia cells
  • Appears as a solid
  • Light yellow to yellow color
  • Purity of 95.80%
Catalog No. 50-003-72703 Supplier Medchemexpress LLC Supplier No. HY1534695MG
Benzyl DC-81 (Compound 6a) is an anticancer agent with antiproliferative activity against A375 and MCF-7 cells. It has an IC50 of 1 μM against CH1 human ovarian cells and 3 μM against L1210 Leukemia cells. This product is intended for research use only.
  • Anticancer agent
  • Antiproliferative activity against A375 and MCF-7 cells
  • IC50 of 1 μM against CH1 human ovarian cells
  • IC50 of 3 μM against L1210 Leukemia cells
  • Appears as a solid
  • Light yellow to yellow color
  • Purity of 95.80%