Learn More
Medchemexpress LLC Benzyl DC-81 | 127810-79-1 | MFCD03404776 | 95.8% | 336.38 | C20H20N2O3 | 10 MG
SDP
Catalog No. 5000286928
Encompass_Preferred
Benzyl DC-81 is an investigational anticancer small molecule reported to inhibit proliferation of A375 (melanoma) and MCF-7 (breast cancer) cell lines. It is supplied for research and drug-registration application purposes and is provided with batch-specific analytical documentation.
  • Reported antiproliferative activity against A375 and MCF-7 cells.
  • Supplied with batch certificate of analysis and purity data.
  • Available in multiple pack sizes and a DMSO stock solution for screening.
  • Characterized by molecular formula and molecular weight for compound confirmation.
  • Intended for in vitro and preclinical research use only.
Catalog No. 50-002-86928 Supplier Medchemexpress LLC Supplier No. HY15346910MG
Benzyl DC-81 is an investigational anticancer small molecule reported to inhibit proliferation of A375 (melanoma) and MCF-7 (breast cancer) cell lines. It is supplied for research and drug-registration application purposes and is provided with batch-specific analytical documentation.
  • Reported antiproliferative activity against A375 and MCF-7 cells.
  • Supplied with batch certificate of analysis and purity data.
  • Available in multiple pack sizes and a DMSO stock solution for screening.
  • Characterized by molecular formula and molecular weight for compound confirmation.
  • Intended for in vitro and preclinical research use only.