Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzyl DL-Mandelate 98.0+%, TCI America™
SDP

Catalog No. M0711500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M0711500G 500 g
M071125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M0711500G Supplier TCI America Supplier No. M0711500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 890-98-2
Molecular Formula C15H14O3
Molecular Weight (g/mol) 242.274
MDL Number MFCD00021856
InChI Key JFKWZVQEMSKSBU-UHFFFAOYSA-N
Synonym DL-Mandelic Acid Benzyl Ester
PubChem CID 13461
IUPAC Name benzyl 2-hydroxy-2-phenylacetate
SMILES C1=CC=C(C=C1)COC(=O)C(C2=CC=CC=C2)O

Specifications

Melting Point 95°C
Color White
Quantity 500 g
Formula Weight 242.27
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Benzyl DL-Mandelate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.