Learn More
Medchemexpress LLC Benzyl-α-GalNAc | 3554-93-6 | 99.9% | 1 ML
SDP
Catalog No. 5000342304
Encompass_Preferred
Benzyl-α-GalNAc is a potent inhibitor of O-glycosylation. It effectively inhibits the proliferation and activation of LX-2 cells and suppresses the expression of collagen I/III, indicating its potential for investigating liver fibrosis. It also significantly enhances the anti-tumor activity of 5-Fluorouracil (e.g., in pancreatic cancer) by inhibiting O-glycosylation.
  • Potent O-glycosylation inhibitor
  • Inhibits proliferation and activation of LX-2 cells
  • Suppresses expression of collagen I/III
  • Investigates liver fibrosis
  • Enhances anti-tumor activity of 5-Fluorouracil
Catalog No. 50-003-42304 Supplier Medchemexpress LLC Supplier No. HY12938910MM/1ML
Benzyl-α-GalNAc is a potent inhibitor of O-glycosylation. It effectively inhibits the proliferation and activation of LX-2 cells and suppresses the expression of collagen I/III, indicating its potential for investigating liver fibrosis. It also significantly enhances the anti-tumor activity of 5-Fluorouracil (e.g., in pancreatic cancer) by inhibiting O-glycosylation.
  • Potent O-glycosylation inhibitor
  • Inhibits proliferation and activation of LX-2 cells
  • Suppresses expression of collagen I/III
  • Investigates liver fibrosis
  • Enhances anti-tumor activity of 5-Fluorouracil