Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Benzyl glycidyl ether

Catalog No. 4697855ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
46-978-55ML 5 mL
46-978-525ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 46-978-55ML Supplier Sigma Aldrich Supplier No. 4697855ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Benzyl glycidyl ether is a glycidyl derivative
  • Rac-/(r)-benzyl glycidyl ether participates in the synthesis of atactic and isotactic linear poly(benzyl 1,2-glycerol carbonate)s
  • Biocatalytic resolution of racemic benzyl glycidyl ether by talaromyces flavus has been described
  • Resolution of benzyl glycidyl ether to (s)-benzyl glycidyl ether with 30% ee and (r)-3-benzyloxypropane-1,2-diol with 40% ee by whole bacillus alcalophilus cells has been reported
  • It reacts with k-, k+(15-crown-5)2 to afford potassium glycidoxide and bibenzyl

Specifications

Density 1.077 g/mL (at 25°C (literature))
Boiling Point 70°C to 73°C (11 mmHg)
Molecular Formula C10H12O2
Refractive Index n20/D 1.5170 (literature)
Linear Formula C10H12O2
MDL Number MFCD00068664
Quantity 5 mL
Molecular Weight (g/mol) 164.2
Percent Purity 99%
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.