Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzyl Glycidyl Ether 97.0+%, TCI America™
SDP

Catalog No. B21085G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B21085G 5 g
B210825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B21085G Supplier TCI America Supplier No. B21085G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2930-05-4
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.20
MDL Number MFCD00068664
InChI Key QNYBOILAKBSWFG-UHFFFAOYNA-N
Synonym 1-Benzyloxy-2,3-epoxypropane, 2-(Benzyloxymethyl)oxirane
PubChem CID 94247
IUPAC Name 2-(phenylmethoxymethyl)oxirane
SMILES C1C(O1)COCC2=CC=CC=C2

Specifications

Color Yellow
Boiling Point 75°C
Quantity 5 g
Formula Weight 164.20
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Benzyl Glycidyl Ether
RTECSNumber TX2860000
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.