Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Benzyl glycolate

Catalog No. 41092625ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
41-092-625ML 25 mL
41-092-65ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 41-092-625ML Supplier Sigma Aldrich Supplier No. 41092625ML
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Benzyl glycolate is a benzyl ester of glycolic acid.benzyl glycolate may be used for the synthesis of the methyl [2-(2α-benzyloxy-2α-oxoethyl)-5-acetamido-4,7,8,9- tetra-o-acetyl-3,5-dideoxy-α- and -α-d-glycero-d-galacto-2-nonulopyranosid]onate. it may be used for the preparation of monosaccharide building blocks. it may also be used for the synthesis of the following new polymeric phosphazenes having pendent esters of glycolic or lactic acid as side units:
  • Poly[bis(ethyl glycolato)phosphazene]
  • Poly[bis(ethyl lactato)phosphazene]
  • Poly[bis(benzyl glycolato)phosphazene]
  • Poly[bis(benzyl lactato)phosphazene]

Specifications

CAS 30379-58-9
Density 1.171 g/mL (at 25°C (literature))
Boiling Point 136°C (14 mmHg)
Molecular Formula C9H10O3
Refractive Index n20/D 1.527 (literature)
Linear Formula HOCH2CO2CH2C6H5
MDL Number MFCD00191776
Quantity 25 mL
Molecular Weight (g/mol) 166.17
Percent Purity 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.