Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzyl Isobutyrate 98.0+%, TCI America™
SDP

Catalog No. I042425ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I042425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I042425ML Supplier TCI America Supplier No. I042425ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 103-28-6
Molecular Formula C11H14O2
Molecular Weight (g/mol) 178.231
MDL Number MFCD00048315
InChI Key UIKJRDSCEYGECG-UHFFFAOYSA-N
Synonym benzyl isobutyrate, benzyl isobutanoate, benzyl 2-methylpropionate, phenylmethyl 2-methylpropanoate, isobutyric acid, benzyl ester, propanoic acid, 2-methyl-, phenylmethyl ester, benzyl 2-methyl propionate, benzylester kyseliny isomaselne, unii-p98pe45v9m, benzyl isobutyrate natural
PubChem CID 7646
IUPAC Name benzyl 2-methylpropanoate
SMILES CC(C)C(=O)OCC1=CC=CC=C1

Specifications

Color Colorless
Boiling Point 230°C
Quantity 25 mL
Formula Weight 178.23
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Benzyl Isobutyrate
EINECSNumber (3)-1020&(3)-1045
RTECSNumber NQ4550000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.