Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzyl Isopropenyl Ether 97.0+%, TCI America™
SDP

Catalog No. B12575ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B12575ML 5 mL
B12571ML 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B12575ML Supplier TCI America Supplier No. B12575ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 32783-20-3
Molecular Formula C10H12O
Molecular Weight (g/mol) 148.205
MDL Number MFCD00059913
InChI Key ZIFHFFOAEFKJJL-UHFFFAOYSA-N
Synonym 2-Benzyloxy-1-propene, Isopropenyl Benzyl Ether
PubChem CID 560989
IUPAC Name prop-1-en-2-yloxymethylbenzene
SMILES CC(=C)OCC1=CC=CC=C1

Specifications

Color Colorless
Boiling Point 192°C
Quantity 5 mL
Formula Weight 148.21
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Benzyl Isopropenyl Ether [Hydroxyl-Protecting Agent]
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.