Learn More
Medchemexpress LLC HY-77813 100mg Medchemexpress, Benzyl isothiocyanate CAS:622-78-6 Purity:>98%
SDP
Catalog No. 501970083
Encompass_Preferred
Medchemexpress, HY-77813 100mg Benzyl isothiocyanate CAS:622-78-6 Benzyl isothiocyanate is a member of natural isothiocyanates with antimicrobial activity. Benzyl isothiocyanate potent inhibits cell mobility, migration and invasion nature and matrix metalloproteinase-2 (MMP-2) activity of murine melanoma cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0083 Supplier Medchemexpress LLC Supplier No. HY77813100MG
Medchemexpress, HY-77813 100mg Benzyl isothiocyanate CAS:622-78-6 Benzyl isothiocyanate is a member of natural isothiocyanates with antimicrobial activity. Benzyl isothiocyanate potent inhibits cell mobility, migration and invasion nature and matrix metalloproteinase-2 (MMP-2) activity of murine melanoma cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.