Learn More
Medchemexpress LLC Benzyl isothiocyanate | 622-78-6 | MFCD00004819 | 99.6% | C8H7NS | 1 ML
SDP
Catalog No. 5000434968
Encompass_Preferred
Benzyl isothiocyanate is an orally available isothiocyanate that shows bactericidal, anticancer, antiangiogenic, and anthelmintic activities. It regulates various signaling pathways including apoptosis, cell proliferation, and angiogenesis. It also enhances muscle insulin sensitivity, improving obesity-induced hyperglycemia.
  • Exhibits bactericidal, anticancer, antiangiogenic, and anthelmintic activities
  • Regulates apoptosis, cell proliferation, cell cycle arrest, metastasis, angiogenesis, and autophagy
  • Enhances muscle insulin sensitivity, improving obesity-induced hyperglycemia
  • Inhibits migration, mobilization, and invasion of B16F10 cells
  • Cytotoxic to *Caenorhabditis elegans* and ovarian tumor cell lines
  • Promotes insulin-dependent glucose uptake in PA-treated C2C12 myotubes
Catalog No. 50-004-34968 Supplier Medchemexpress LLC Supplier No. HY7781310MM/1ML
Benzyl isothiocyanate is an orally available isothiocyanate that shows bactericidal, anticancer, antiangiogenic, and anthelmintic activities. It regulates various signaling pathways including apoptosis, cell proliferation, and angiogenesis. It also enhances muscle insulin sensitivity, improving obesity-induced hyperglycemia.
  • Exhibits bactericidal, anticancer, antiangiogenic, and anthelmintic activities
  • Regulates apoptosis, cell proliferation, cell cycle arrest, metastasis, angiogenesis, and autophagy
  • Enhances muscle insulin sensitivity, improving obesity-induced hyperglycemia
  • Inhibits migration, mobilization, and invasion of B16F10 cells
  • Cytotoxic to *Caenorhabditis elegans* and ovarian tumor cell lines
  • Promotes insulin-dependent glucose uptake in PA-treated C2C12 myotubes