Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Benzyl isothiocyanate

Catalog No. 2524925G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
25-249-25G 5 g
25-249-225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 25-249-25G Supplier Sigma Aldrich Supplier No. 2524925G
May include imposed supplier surcharges.
 Restricted Item
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Benzyl isothiocyanate is a naturally-occurring constituent of cruciferous vegetables
  • It has antibacterial properties and its metabolism in man has been investigated
  • It inhibits chemically induced cancer in animal models.benzyl isothiocyanate has been used in the preparation of s-(n-benzylthiocarbamoyl)-L-glutathione and s-(n-benzylthiocarbamoyl)-L-cysteine

Specifications

CAS 622-78-6
Density 1.125 g/mL (at 25°C (literature))
Boiling Point 242°C to 243°C (lit.)
Molecular Formula C8H7NS
Refractive Index n20/D 1.601 (literature)
Linear Formula C6H5CH2NCS
MDL Number MFCD00004819
Quantity 5 g
Synonym Benzyl mustard oil; Isothiocyanotaomethylbenzene
Molecular Weight (g/mol) 149.21
Percent Purity 98%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.