Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Benzyl methacrylate

Catalog No. 409448250ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
250 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-944-8250ML 250 mL
40-944-81L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 40-944-8250ML Supplier Sigma Aldrich Supplier No. 409448250ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Benzyl methacrylate

Specifications

CAS 2495-37-6
Density 1.04 g/mL (at 25°C)
Boiling Point 95°C to 98°C (4 mmHg)
Molecular Formula C11H12O2
Refractive Index n20/D 1.512 (literature)
Linear Formula H2C=C(CH3)CO2CH2C6H5
Quantity 250 mL
MDL Number MFCD00053684
Synonym Benzyl alpha-methylacrylate; Benzyl 2-methylacrylate; Phenylmethyl 2-methyl-2-propenoate
Molecular Weight (g/mol) 176.21
Percent Purity 96%
Biological Activity Respiratory system
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.