Learn More
Medchemexpress LLC Benzyl N-(2-{2-[2-(2-aminoethoxy)ethoxy]ethoxy}ethyl)carbamate | 863973-20-0 | MFCD28155474 | >98.0% | 326.39 g·mol⁻1 | C16H26N2O5 | 250 MG
SDP
Catalog No. 5000282539
Encompass_Preferred
CbzNH-PEG3-CH2CH2NH2 is a Cbz-protected polyethylene glycol (PEG3) linker with a terminal primary amine. It is supplied as a research building block for organic synthesis and bioconjugation workflows where a short, hydrophilic spacer and a protected amine are required for stepwise coupling and modification.
  • Cbz-protected primary amine for orthogonal deprotection.
  • PEG3 spacer provides hydrophilicity and flexibility.
  • Suitable for linker and conjugate synthesis in medicinal chemistry.
  • Available in small milligram-to-gram quantities for research use.
  • Reported high purity appropriate for synthetic applications.
Catalog No. 50-002-82539 Supplier Medchemexpress LLC Supplier No. HYW348348250MG
CbzNH-PEG3-CH2CH2NH2 is a Cbz-protected polyethylene glycol (PEG3) linker with a terminal primary amine. It is supplied as a research building block for organic synthesis and bioconjugation workflows where a short, hydrophilic spacer and a protected amine are required for stepwise coupling and modification.
  • Cbz-protected primary amine for orthogonal deprotection.
  • PEG3 spacer provides hydrophilicity and flexibility.
  • Suitable for linker and conjugate synthesis in medicinal chemistry.
  • Available in small milligram-to-gram quantities for research use.
  • Reported high purity appropriate for synthetic applications.