Learn More
Medchemexpress LLC Benzyl N-(2-{2-[2-(2-aminoethoxy)ethoxy]ethoxy}ethyl)carbamate | 863973-20-0 | MFCD28155474 | 99.0% | 326.39 g/mol | C16H26N2O5 | 500 MG
SDP
Catalog No. 5000282410
Encompass_Preferred
Benzyl N-(2-{2-[2-(2-aminoethoxy)ethoxy]ethoxy}ethyl)carbamate is a Cbz-protected PEG3 linker bearing a terminal primary amine. The hydrophilic PEG spacer improves aqueous solubility, while the Cbz protecting group enables controlled deprotection, making the reagent useful for installing amine-functional linkers in small-molecule conjugates, bioconjugation workflows, and PROTAC assemblies.
  • Cbz-protected terminal amine for orthogonal deprotection.
  • PEG3 spacer increases water solubility.
  • Primary amine handle for reliable conjugation chemistry.
  • Compatible with linker synthesis and bioconjugation workflows.
  • Available in laboratory-scale quantities for synthesis and screening.
Catalog No. 50-002-82410 Supplier Medchemexpress LLC Supplier No. HYW348348500MG
Add to Cart
Add to Cart
Benzyl N-(2-{2-[2-(2-aminoethoxy)ethoxy]ethoxy}ethyl)carbamate is a Cbz-protected PEG3 linker bearing a terminal primary amine. The hydrophilic PEG spacer improves aqueous solubility, while the Cbz protecting group enables controlled deprotection, making the reagent useful for installing amine-functional linkers in small-molecule conjugates, bioconjugation workflows, and PROTAC assemblies.
  • Cbz-protected terminal amine for orthogonal deprotection.
  • PEG3 spacer increases water solubility.
  • Primary amine handle for reliable conjugation chemistry.
  • Compatible with linker synthesis and bioconjugation workflows.
  • Available in laboratory-scale quantities for synthesis and screening.