Learn More
Medchemexpress LLC Benzyl N-{2-[2-(2-bromoethoxy)ethoxy]ethyl}carbamate | 2100283-00-7 | MFCD31916729 | 97.0% | 346.22 g/mol | C14H20BrNO4 | 100 MG
SDP
Catalog No. 5000280775
Encompass_Preferred
Cbz-PEG2-bromide is a carbobenzyloxy (Cbz)-protected PEG2 linker bearing a terminal bromide functional group. It is supplied as a high-purity synthetic building block for use in the assembly of PROTACs and other heterobifunctional molecules where a short polyethylene glycol spacer and an alkylation handle are required.
  • Cbz-protected amine compatible with common deprotection conditions.
  • Terminal bromide enables alkylation and nucleophilic substitution chemistry.
  • Short PEG2 spacer provides flexibility and improved solubility in organic solvents.
  • High purity suitable for synthetic and medicinal chemistry applications.
  • Available in multiple pack sizes to support small-scale synthesis.
Catalog No. 50-002-80775 Supplier Medchemexpress LLC Supplier No. HYW190968100MG
Cbz-PEG2-bromide is a carbobenzyloxy (Cbz)-protected PEG2 linker bearing a terminal bromide functional group. It is supplied as a high-purity synthetic building block for use in the assembly of PROTACs and other heterobifunctional molecules where a short polyethylene glycol spacer and an alkylation handle are required.
  • Cbz-protected amine compatible with common deprotection conditions.
  • Terminal bromide enables alkylation and nucleophilic substitution chemistry.
  • Short PEG2 spacer provides flexibility and improved solubility in organic solvents.
  • High purity suitable for synthetic and medicinal chemistry applications.
  • Available in multiple pack sizes to support small-scale synthesis.