Learn More
Medchemexpress LLC Benzyl N-[(2S)-1-hydroxy-3-phenylpropan-2-yl]carbamate | 6372-14-1 | >98.0% | 285.34 g·mol⁻¹ | C17H19NO3 | 10 G
SDP
Catalog No. 5000303837
Encompass_Preferred
Z-Phenylalaninol is a Cbz-protected phenylalaninol (N-benzyloxycarbonyl-L-phenylalaninol) used as an amino-alcohol building block and intermediate in organic synthesis, peptide chemistry, and stereoselective transformations. It is supplied as a solid powder (CAS 6372-14-1) with molecular formula C17H19NO3 and molecular weight 285.34 g·mol⁻¹.
Catalog No. 50-003-03837 Supplier Medchemexpress LLC Supplier No. HYW01384410G
Z-Phenylalaninol is a Cbz-protected phenylalaninol (N-benzyloxycarbonyl-L-phenylalaninol) used as an amino-alcohol building block and intermediate in organic synthesis, peptide chemistry, and stereoselective transformations. It is supplied as a solid powder (CAS 6372-14-1) with molecular formula C17H19NO3 and molecular weight 285.34 g·mol⁻¹.