Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Benzyl N-[(2S,3S)-3-methyl-1-[[(2S)-4-methyl-1-oxopentan-2-yl]amino]-1-oxopentan-2-yl]carbamate | 161710-10-7 | MFCD04974201 | 97.3% | 362.46 g/mol | C20H30N2O4 | 1MG
SDP

Catalog No. 5000236596
Encompass_Preferred

Z-Ile-Leu-aldehyde (Z-IL-CHO) is a peptide aldehyde inhibitor that competitively inhibits γ-secretase and Notch signaling. It is supplied for research use as a solid or as a 10 mM solution in DMSO and is used to investigate γ-secretase activity, Notch pathway modulation, and related cellular processes.

  • Peptide aldehyde inhibitor of γ-secretase and Notch.
  • Chemical formula C20H30N2O4; molecular weight 362.46 g/mol.
  • Purity 97.3% by HPLC.
  • Available as solid and as 10 mM solution in DMSO.
  • Storage: powder -20 °C (3 years), 4 °C (2 years); in solvent -80 °C (6 months), -20 °C (1 month).
  • Intended for research use only.

Catalog No. 50-002-36596 Supplier Medchemexpress LLC Supplier No. HY124651MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Z-Ile-Leu-aldehyde (Z-IL-CHO) is a peptide aldehyde inhibitor that competitively inhibits γ-secretase and Notch signaling. It is supplied for research use as a solid or as a 10 mM solution in DMSO and is used to investigate γ-secretase activity, Notch pathway modulation, and related cellular processes.

  • Peptide aldehyde inhibitor of γ-secretase and Notch.
  • Chemical formula C20H30N2O4; molecular weight 362.46 g/mol.
  • Purity 97.3% by HPLC.
  • Available as solid and as 10 mM solution in DMSO.
  • Storage: powder -20 °C (3 years), 4 °C (2 years); in solvent -80 °C (6 months), -20 °C (1 month).
  • Intended for research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.