Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Benzyl N6-(t-Boc)-L-lysinate | 133170-57-7 | MFCD11045868 | 97.5% | 372.89 | C18H29ClN2O4 | 10 G
SDP

Catalog No. 5000208549
Encompass_Preferred

Benzyl N6-(t-Boc)-L-lysinate is a Boc-protected benzyl derivative of L-lysine used as a reagent and building block in peptide synthesis and organic chemistry research. It is supplied for laboratory research use only; handle according to standard safety procedures.

  • Boc-protected lysine derivative.
  • Cas 133170-57-7.
  • Molecular formula C18H29ClN2O4.
  • Molecular weight 372.89.
  • Purity approximately 97.5% (LCMS).
  • Storage: store dry at room temperature; in solvent store -80°C (2 years) or -20°C (1 year).

Catalog No. 50-002-08549 Supplier Medchemexpress LLC Supplier No. HYW08871210G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Benzyl N6-(t-Boc)-L-lysinate is a Boc-protected benzyl derivative of L-lysine used as a reagent and building block in peptide synthesis and organic chemistry research. It is supplied for laboratory research use only; handle according to standard safety procedures.

  • Boc-protected lysine derivative.
  • Cas 133170-57-7.
  • Molecular formula C18H29ClN2O4.
  • Molecular weight 372.89.
  • Purity approximately 97.5% (LCMS).
  • Storage: store dry at room temperature; in solvent store -80°C (2 years) or -20°C (1 year).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.