Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzyl Orange, TCI America™
SDP

Catalog No. B04251G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B04251G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B04251G Supplier TCI America Supplier No. B04251G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 36402-77-4
Molecular Formula C19H16N3NaO3S
Molecular Weight (g/mol) 389.405
MDL Number MFCD00198097
InChI Key JGCBNZLRTUKUAQ-UHFFFAOYSA-M
PubChem CID 60161325
IUPAC Name sodium;4-[[4-(benzylamino)phenyl]diazenyl]benzenesulfonate
SMILES C1=CC=C(C=C1)CNC2=CC=C(C=C2)N=NC3=CC=C(C=C3)S(=O)(=O)[O-].[Na+]

Specifications

Quantity 1 g
Formula Weight 389.40
Physical Form Crystalline Powder
Chemical Name or Material Benzyl Orange
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.