Learn More
Medchemexpress LLC Benzyl Paraben-d7 | 1216916-40-3 | 235.29 | 5 MG
SDP
Catalog No. 5000454266
Encompass_Preferred
Benzyl Paraben-d7 is a deuterium-labeled Benzyl 4-hydroxybenzoate, which is a prominent material used as an excipient. It can function as a bacteriostatic agent or preservative in pharmaceutical preparations, improving stability, solubility, and processability. Excipients also influence the absorption, distribution, metabolism, and elimination (ADME) processes of co-administered drugs. This product is for research use only and has not been fully validated for medical applications.
  • Deuterium-labeled compound
  • Can be used as a bacteriostatic agent
  • Functions as a preservative
  • Improves stability and solubility of pharmaceutical preparations
  • Affects ADME processes of co-administered drugs
Catalog No. 50-004-54266 Supplier Medchemexpress LLC Supplier No. HYW013482S15MG
Add to Cart
Add to Cart
Benzyl Paraben-d7 is a deuterium-labeled Benzyl 4-hydroxybenzoate, which is a prominent material used as an excipient. It can function as a bacteriostatic agent or preservative in pharmaceutical preparations, improving stability, solubility, and processability. Excipients also influence the absorption, distribution, metabolism, and elimination (ADME) processes of co-administered drugs. This product is for research use only and has not been fully validated for medical applications.
  • Deuterium-labeled compound
  • Can be used as a bacteriostatic agent
  • Functions as a preservative
  • Improves stability and solubility of pharmaceutical preparations
  • Affects ADME processes of co-administered drugs