Learn More
Medchemexpress LLC Benzyl-PEG10-alcohol 50mg
SDP
Catalog No. 5000206030
Encompass_Preferred
Benzyl-PEG10-alcohol is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs[1]
Catalog No. 50-002-06030 Supplier Medchemexpress LLC Supplier No. HYW09608150MG
Benzyl-PEG10-alcohol is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs[1]