Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Benzyl-PEG2-amine | 1268135-96-1 | >97.7% | C11H17NO2 | 100 MG
SDP

Catalog No. 5000353358
Encompass_Preferred

Benzyl-PEG2-amine is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. These PROTACs facilitate the connection of ligands for E3 ubiquitin ligase and target proteins, enabling the degradation of target proteins via the ubiquitin-proteasome system.

  • Functions as a PEG-based linker for PROTACs.
  • Utilized in the synthesis of PROTACs.
  • Molecular weight: 195.26.
  • Chemical formula: C11H17NO2.
  • Appearance: Colorless to light yellow liquid.
  • Storage: Recommended at 4°C, protected from light; in solvent, -80°C for 6 months or -20°C for 1 month, protected from light.
  • Enables degradation of target proteins via the ubiquitin-proteasome system.

Catalog No. 50-003-53358 Supplier Medchemexpress LLC Supplier No. HY140742100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Benzyl-PEG2-amine is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. These PROTACs facilitate the connection of ligands for E3 ubiquitin ligase and target proteins, enabling the degradation of target proteins via the ubiquitin-proteasome system.

  • Functions as a PEG-based linker for PROTACs.
  • Utilized in the synthesis of PROTACs.
  • Molecular weight: 195.26.
  • Chemical formula: C11H17NO2.
  • Appearance: Colorless to light yellow liquid.
  • Storage: Recommended at 4°C, protected from light; in solvent, -80°C for 6 months or -20°C for 1 month, protected from light.
  • Enables degradation of target proteins via the ubiquitin-proteasome system.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.