Learn More
Medchemexpress LLC Benzyl-PEG2-amine | 1268135-96-1 | C11H17NO2 | 1 G
SDP
Catalog No. 5000352962
Encompass_Preferred
Benzyl-PEG2-amine is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Connects ligands for E3 ubiquitin ligase and target proteins
  • Utilizes the ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-52962 Supplier Medchemexpress LLC Supplier No. HY1407421G
Benzyl-PEG2-amine is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Connects ligands for E3 ubiquitin ligase and target proteins
  • Utilizes the ubiquitin-proteasome system
  • Selectively degrades target proteins