Learn More
Medchemexpress LLC Benzyl-PEG2-amine | 1268135-96-1 | C11H17NO2 | 250 MG
SDP
Catalog No. 5000352555
Encompass_Preferred
Benzyl-PEG2-amine is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs are designed with two distinct ligands linked together; one ligand binds to an E3 ubiquitin ligase and the other targets a specific protein. This mechanism allows PROTACs to leverage the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Functions as a PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Facilitates targeted protein degradation
  • PROTACs utilize specific ligands for E3 ubiquitin ligase and target proteins
Catalog No. 50-003-52555 Supplier Medchemexpress LLC Supplier No. 250MGHY140742
Benzyl-PEG2-amine is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs are designed with two distinct ligands linked together; one ligand binds to an E3 ubiquitin ligase and the other targets a specific protein. This mechanism allows PROTACs to leverage the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Functions as a PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Facilitates targeted protein degradation
  • PROTACs utilize specific ligands for E3 ubiquitin ligase and target proteins