Learn More
Medchemexpress LLC Benzyl-PEG2-azide | 1260001-87-3 | 99.1% | 100 MG
SDP
Catalog No. 5000339619
Encompass_Preferred
Benzyl-PEG2-azide is a PEG-based PROTAC linker used in the synthesis of PROTACs. It functions as a click chemistry reagent due to its azide group, enabling copper-catalyzed azide-alkyne cycloaddition (CuAAc) with alkyne-containing molecules, and strain-promoted alkyne-azide cycloaddition (SPAAC) with DBCO or BCN groups.
  • Functions as a PEG-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • Acts as a click chemistry reagent with an azide group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) with alkyne groups
  • Allows for strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with DBCO or BCN groups
Catalog No. 50-003-39619 Supplier Medchemexpress LLC Supplier No. HY130188100MG
Benzyl-PEG2-azide is a PEG-based PROTAC linker used in the synthesis of PROTACs. It functions as a click chemistry reagent due to its azide group, enabling copper-catalyzed azide-alkyne cycloaddition (CuAAc) with alkyne-containing molecules, and strain-promoted alkyne-azide cycloaddition (SPAAC) with DBCO or BCN groups.
  • Functions as a PEG-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • Acts as a click chemistry reagent with an azide group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) with alkyne groups
  • Allows for strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with DBCO or BCN groups