Learn More
Medchemexpress LLC Benzyl-PEG2-CH2-Boc | 1643957-26-9 | 98.0% | 1 G
SDP
Catalog No. 5000339647
Encompass_Preferred
Benzyl-PEG2-CH2-Boc is a PEG-based PROTAC linker that can be utilized in the synthesis of PROTACs. PROTACs consist of two distinct ligands connected by a linker, with one ligand targeting an E3 ubiquitin ligase and the other targeting a protein of interest. PROTACs function by leveraging the intracellular ubiquitin-proteasome system to selectively degrade target proteins. This product is for research use only.
  • PEG-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • PROTACs consist of two distinct ligands connected by a linker
  • PROTACs leverage the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-39647 Supplier Medchemexpress LLC Supplier No. HY1301721G
Benzyl-PEG2-CH2-Boc is a PEG-based PROTAC linker that can be utilized in the synthesis of PROTACs. PROTACs consist of two distinct ligands connected by a linker, with one ligand targeting an E3 ubiquitin ligase and the other targeting a protein of interest. PROTACs function by leveraging the intracellular ubiquitin-proteasome system to selectively degrade target proteins. This product is for research use only.
  • PEG-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • PROTACs consist of two distinct ligands connected by a linker
  • PROTACs leverage the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins