Learn More
Medchemexpress LLC Benzyl-PEG4-Boc | 2484091-04-3 | 368.46 | 250 MG
SDP
Catalog No. 5000348250
Encompass_Preferred
Benzyl-PEG4-Boc is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs are designed with two different ligands connected by a linker, one for an E3 ubiquitin ligase and another for the target protein. They harness the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Facilitates selective degradation of target proteins
Catalog No. 50-003-48250 Supplier Medchemexpress LLC Supplier No. 250MGHY138346
Benzyl-PEG4-Boc is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs are designed with two different ligands connected by a linker, one for an E3 ubiquitin ligase and another for the target protein. They harness the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Facilitates selective degradation of target proteins