Learn More
Medchemexpress LLC Benzyl-PEG4-Ots 500mg
SDP
Catalog No. 5000206706
Encompass_Preferred
Benzyl-PEG4-Ots is a PEG-based PROTAC linker can be used in the synthesis of PROTACs
Catalog No. 50-002-06706 Supplier Medchemexpress LLC Supplier No. HYW108219500MG
Benzyl-PEG4-Ots is a PEG-based PROTAC linker can be used in the synthesis of PROTACs