Learn More
Medchemexpress LLC Benzyl-PEG45-alcohol | 26403-74-7 | C97H188O46 | 100 MG
SDP
Catalog No. 5000373443
Encompass_Preferred
Benzyl-PEG45-alcohol is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PROTACs contain two different ligands connected by a linker.
  • One ligand binds to an E3 ubiquitin ligase and the other to the target protein.
  • Enables selective degradation of target proteins.
Catalog No. 50-003-73443 Supplier Medchemexpress LLC Supplier No. HY153153100MG
Benzyl-PEG45-alcohol is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PROTACs contain two different ligands connected by a linker.
  • One ligand binds to an E3 ubiquitin ligase and the other to the target protein.
  • Enables selective degradation of target proteins.