Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Benzyl-PEG5-amine | 86770-77-6 | 98.0% | C17H29NO5 | 1 G
SDP

Catalog No. 5000353792
Encompass_Preferred

Benzyl-PEG5-amine is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins. It has a molecular weight of 327.42, appears as a liquid, and is colorless to light yellow.

  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • Molecular weight of 327.42
  • Liquid appearance
  • Colorless to light yellow
  • Purity of 98.0%

Catalog No. 50-003-53792 Supplier Medchemexpress LLC Supplier No. HY1407431G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Benzyl-PEG5-amine is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins. It has a molecular weight of 327.42, appears as a liquid, and is colorless to light yellow.

  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • Molecular weight of 327.42
  • Liquid appearance
  • Colorless to light yellow
  • Purity of 98.0%

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.