Learn More
Medchemexpress LLC Benzyl-PEG5-amine | 86770-77-6 | 98.0% | C17H29NO5 | 25 MG
SDP
Catalog No. 5000352514
Encompass_Preferred
Benzyl-PEG5-amine is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs are molecules with two ligands connected by a linker, one binding to an E3 ubiquitin ligase and the other to a target protein. This mechanism exploits the intracellular ubiquitin-proteasome system for selective degradation of target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Contains two different ligands connected by a linker
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-52514 Supplier Medchemexpress LLC Supplier No. 25MGHY140743
Benzyl-PEG5-amine is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs are molecules with two ligands connected by a linker, one binding to an E3 ubiquitin ligase and the other to a target protein. This mechanism exploits the intracellular ubiquitin-proteasome system for selective degradation of target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Contains two different ligands connected by a linker
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins